missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxane-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 11419117
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419117 5 g 5g
11409117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419117 Supplier Thermo Scientific Alfa Aesar Supplier No. B20661.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3663-80-7
Molecular Formula C9H7O4
Molecular Weight (g/mol) 179.15
MDL Number MFCD00084888
InChI Key HMBHAQMOBKLWRX-QMMMGPOBSA-M
Synonym 1,4-benzodioxan-2-carboxylic acid, 1,4-benzodioxane-2-carboxylic acid, 2,3-dihydro-1,4-benzodioxine-2-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-2-carboxylic acid, 1,4-benzodioxin-2-carboxylic acid, 2,3-dihydro, 1,4-benzodioxan-2-carboxylicacid, 2rs-2,3-dihydro-1,4-benzodioxine-2-carboxylic acid, rac 1,4-benzodioxane-2-carboxylic acid, 2,3-dihydrobenzo 1,4 dioxine-2-carboxylic acid
PubChem CID 2735450
IUPAC Name 2,3-dihydro-1,4-benzodioxine-3-carboxylic acid
SMILES [O-]C(=O)[C@@H]1COC2=CC=CC=C2O1

Tekniske data

Melting Point 122°C to 126°C
Quantity 5 g
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 1,4-Benzodioxane-2-carboxylic acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.