missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Benzodioxan, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 493-09-4 |
---|---|
Molecular Formula | C8H8O2 |
Molecular Weight (g/mol) | 136.15 |
MDL Number | MFCD00006821 |
InChI Key | BNBQRQQYDMDJAH-UHFFFAOYSA-N |
Synonym | 1,4-benzodioxane, 1,4-benzodioxan, 2,3-dihydrobenzo b 1,4 dioxine, 1,4-benzodioxin, 2,3-dihydro, pyrocatechol ethylene ether, 1,2-ethylenedioxy benzene, benzodioxan, ethylene o-phenylene dioxide, 1,2-ethylenedioxybenzene, 1, 4-benzdioxan |
PubChem CID | 10301 |
IUPAC Name | 2,3-dihydro-1,4-benzodioxine |
SMILES | C1COC2=CC=CC=C2O1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10665052
|
Thermo Scientific Acros
176730250 |
25 mL | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
493-09-4 | |
136.15 | |
BNBQRQQYDMDJAH-UHFFFAOYSA-N | |
10301 | |
C1COC2=CC=CC=C2O1 |
C8H8O2 | |
MFCD00006821 | |
1,4-benzodioxane, 1,4-benzodioxan, 2,3-dihydrobenzo b 1,4 dioxine, 1,4-benzodioxin, 2,3-dihydro, pyrocatechol ethylene ether, 1,2-ethylenedioxy benzene, benzodioxan, ethylene o-phenylene dioxide, 1,2-ethylenedioxybenzene, 1, 4-benzdioxan | |
2,3-dihydro-1,4-benzodioxine |
Specifications
493-09-4 | |
100.0 | |
1.1400g/mL | |
87°C | |
98.5% min. (GC) | |
1.5485 to 1.5505 | |
19, 22 | |
1,4-benzodioxane, 1,4-benzodioxan, 2,3-dihydrobenzo b 1,4 dioxine, 1,4-benzodioxin, 2,3-dihydro, pyrocatechol ethylene ether, 1,2-ethylenedioxy benzene, benzodioxan, ethylene o-phenylene dioxide, 1,2-ethylenedioxybenzene, 1, 4-benzdioxan | |
BNBQRQQYDMDJAH-UHFFFAOYSA-N | |
2,3-dihydro-1,4-benzodioxine | |
10301 | |
99% | |
1, 4-Benzodioxan |
98.5 | |
Colorless to Yellow | |
248°C | |
Authentic | |
C8H8O2 | |
MFCD00006821 | |
1.14 | |
Solubility in water: insoluble. Other solubilities: soluble in all typical organic solvents | |
C1COC2=CC=CC=C2O1 | |
136.15 | |
136.15 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 207-775-6