missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-6-yl methyl ketone, 98%, Thermo Scientific™

Product Code. 10299823 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10299823 50 g Glass bottle 50g
10205900 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10299823 Supplier Thermo Scientific Acros Supplier No. 176750500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2879-20-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00006823
InChI Key HGVWMTAIIYNQSI-UHFFFAOYSA-N
PubChem CID 76143
IUPAC Name 1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
SMILES CC(=O)C1=CC=C2OCCOC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 81°C to 85°C
Color Blue to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 50 g
Synonym 6-acetyl-1,4-benzodioxane, 1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone, 1,4-benzodioxan-6-yl methyl ketone, 1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one, 6-acetyl-1,4-benzodioxan, ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl, ethanone,1-2,3-dihydro-1,4-benzodioxin-6-yl, 1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethan-1-one, 6-acetyl-2h,3h-benzo e 1,4-dioxin
Formula Weight 178.19
Percent Purity 98%
Physical Form Fine Crystalline Needles
Chemical Name or Material 1, 4-Benzodioxan-6-yl methyl ketone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.