missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenedimethanol, 99%

Product Code. 11489090
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489090 25 g 25g
11479090 5 g 5g
11499090 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489090 Supplier Thermo Scientific Alfa Aesar Supplier No. A14405.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 589-29-7 | C8H10O2 | 138.166 g/mol

1,4-Benzenedimethanol is used as an organic synthesis intermediate. It is also used for preparation of soluble polyphenylene and p-tolyl-methanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Benzenedimethanol is used as an organic synthesis intermediate. It is also used for preparation of soluble polyphenylene and p-tolyl-methanol.

Solubility
Soluble in ether, toluene, methanol and hot water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-29-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004665
InChI Key BWVAOONFBYYRHY-UHFFFAOYSA-N
Synonym 1,4-benzenedimethanol, p-xylylene glycol, p-xylene glycol, terephthalyl alcohol, 4-hydroxymethyl phenyl methanol, 1,4-xylylene glycol, 1,4-phenylenedimethanol, p-benzenedimethanol, p-xylylenediol, 1,4-dimethylolbenzene
PubChem CID 11506
IUPAC Name [4-(hydroxymethyl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)CO

Specifications

Melting Point 117°C to 121°C
Boiling Point 138°C to 143°C (1 mmHg)
Quantity 25 g
Beilstein 2042077
Solubility Information Soluble in ether,toluene,methanol and hot water.
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material 1,4-Benzenedimethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.