missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenediboronic acid, 96%

Product Code. 11471047
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471047 1 g 1g
11481047 5 g 5g
11491047 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471047 Supplier Thermo Scientific Alfa Aesar Supplier No. B24064.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4612-26-4 | C6H8B2O4 | 165.746 g/mol

It is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.

Solubility
Soluble in water 2.5%.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4612-26-4
Molecular Formula C6H8B2O4
Molecular Weight (g/mol) 165.746
MDL Number MFCD00236018
InChI Key BODYVHJTUHHINQ-UHFFFAOYSA-N
Synonym 1,4-benzenediboronic acid, 1,4-phenylenediboronic acid, 1,4-phenylenebisboronic acid, benzene-1,4-diboronic acid, 4-boronophenyl boronic acid, p-benzenediboronic acid, p-phenylenediboronic acid, 1,4-phenylenebis boronic acid, 1,4 phenyl diboronic acid, boronic acid, 1,4-phenylenebis
PubChem CID 230478
IUPAC Name (4-boronophenyl)boronic acid
SMILES B(C1=CC=C(C=C1)B(O)O)(O)O

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 2836921
Solubility Information Soluble in water 2.5%.
Formula Weight 165.75
Percent Purity 96%
Chemical Name or Material 1,4-Benzenediboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.