missing translation for 'onlineSavingsMsg'
Learn More

1,4-Anhydro-D-sorbitol, TRC

Product Code. 18050620
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18050620 5 g 5g
18083700 1 g 1g
18010620 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18050620 Supplier Toronto Research Chemicals Supplier No. TRCA6555255G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27299-12-3
Molecular Formula C6 H12 O5
Molecular Weight (g/mol) 164.16
Synonym 1,4-Anhydro-D-glucitol, Arlitan (6CI), Glucitol, 1,4-anhydro-, D- (8CI), 1,4-Anhydro-D-glucitol, 1,4-Anhydro-D-sorbitol, 1,4-Anhydroglucitol, 1,4-Sorbitan
IUPAC Name (2R,3R,4S)-2-[(1R)-1,2-dihydroxyethyl]oxolane-3,4-diol
SMILES OC[C@@H](O)[C@H]1OC[C@H](O)[C@H]1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H12O5/c7-1-3(8)6-5(10)4(9)2-11-6/h3-10H,1-2H2/t3-,4+,5-,6-/m1/s1
Formula Weight 164.0685
Percent Purity >95%
Chemical Name or Material 1,4-Sorbitan
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.