missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Amino-2-n-propyl-5-pyrimidinylmethyl)-2-methylpyridinium chloride, Thermo Scientific™

Product Code. 15490387
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490387 25 g 25g
15400397 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490387 Supplier Thermo Scientific Alfa Aesar Supplier No. J64782.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A thiamine analogue; blocks thiamine uptake and prevents carbohydrate synthesis

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-88-2
Molecular Formula C14H20Cl2N4
Molecular Weight (g/mol) 315.24
MDL Number MFCD00078831
InChI Key PJBQYZZKGNOKNJ-UHFFFAOYSA-M
Synonym amprolium hydrochloride, amprolium hcl, amprolium, mepyrium, amprol, amprol 25, thiacoccid, amprol 25 per cent, amprolsol vet, corid vet
PubChem CID 8732
IUPAC Name 5-[(2-methylpyridin-1-ium-1-yl)methyl]-2-propylpyrimidin-4-amine;chloride;hydrochloride
SMILES [H+].[Cl-].[Cl-].CCCC1=NC=C(C[N+]2=CC=CC=C2C)C(N)=N1

Specifications

Color White
Quantity 25 g
Beilstein 4118699
Formula Weight 315.24
Physical Form Powder
Chemical Name or Material 1-(4-Amino-2-n-propyl-5-pyrimidinylmethyl)-2-methylpyridinium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.