missing translation for 'onlineSavingsMsg'
Learn More

1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane

Product Code. 15466065
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466065 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15466065 Supplier Thermo Scientific Alfa Aesar Supplier No. 030854.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41203-22-9 | C14H36N4 | 260.47 g/mol

1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane is used as a macrocyclic chelating agent in coordination chemistry. It is also used as a ligand which bind strongly to a wide range of metal ions and as crown ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 41203-22-9
Melting Point 34°C to 36°C
Flash Point >110°C (230°F)
Molecular Formula C14H36N4
MDL Number MFCD00005106
Quantity 1 g
Sensitivity Hygroscopic
Synonym 1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane, tetramethylcyclam, 1,4,8,11-tetraazacyclotetradecane, 1,4,8,11-tetramethyl, acmc-1ankx, 1,4,8,11-tetramethylcyclam, 1,4,8,11-tetraazacyclotetradecane,1,4,8,11-tetramethyl, 1,4,8,11-tetramethyl-1,4,8, 11-tetraazacyclotetradecane, 1r,4s,8r,11s-1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane, 1alpha,4beta,8beta,11alpha-tetramethyl-1,4,8,11-tetraazacyclotetradecane
Solubility Information Soluble in water.
InChI Key HRFJEOWVAGSJNW-UHFFFAOYNA-R
SMILES C[NH+]1CCC[NH+](C)CC[NH+](C)CCC[NH+](C)CC1
IUPAC Name 1,4,8,11-tetramethyl-1,4,8,11-tetrazacyclotetradecane
Molecular Weight (g/mol) 260.47
PubChem CID 123502
Formula Weight 256.43
Physical Form Crystalline
Chemical Name or Material 1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.