missing translation for 'onlineSavingsMsg'
Learn More

1,4,8,11-Tetraazacyclotetradecane, 98+%

Product Code. p-7081514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444513 1 g 1g
11454513 5 g 5g
11464513 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444513 Supplier Thermo Scientific Alfa Aesar Supplier No. A11516.03
Add to basket
This item is not returnable. View return policy

CAS: 295-37-4 | C10H24N4 | 200.33 g/mol

1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 295-37-4
Molecular Formula C10H24N4
Molecular Weight (g/mol) 200.33
MDL Number MFCD00005105
InChI Key MDAXKAUIABOHTD-UHFFFAOYSA-N
Synonym 1,4,8,11-tetraazacyclotetradecane, cyclam, 14 anen4, 1,4,8,11-tetraazacyclo-tetradecane, 1,4,8,11-tetraazacyclotetradecane cyclam, ips2, acmc-209h9n, 5-26-11-00027 beilstein handbook reference, 1,8,11-tetraazacyclotetradecane, 1,4,8,11tetraaza-cyclotetradecane
PubChem CID 64964
ChEBI CHEBI:37401
IUPAC Name 1,4,8,11-tetrazacyclotetradecane
SMILES C1CNCCNCCCNCCNC1

Specifications

Melting Point 184°C to 188°C
Quantity 1 g
Beilstein 111811
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water.
Formula Weight 200.33
Percent Purity ≥98%
Chemical Name or Material 1,4,8,11-Tetraazacyclotetradecane
compliance-icons
Product Identifier
  • 1,4,8,11-Tetraazacyclotetradecane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,4,8,11-Tetraazacyclotetradecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.