missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1,4,7-Tris(ethoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
Specifications
Specifications
| CAS | 114873-52-8 |
| Molecular Formula | C20H38N4O6 |
| MDL Number | MFCD09263320 |
| Quantity | 250 mg |
| Synonym | triethyl 2,2',2-1,4,7,10-tetraazacyclododecane-1,4,7-triyl triacetate, 1,4,7-tris ethoxycarbonylmethyl-1,4,7,10-tetraazacyclododecane, ethyl 2-4,7-bis 2-ethoxy-2-oxoethyl-1,4,7,10-tetraazacyclododecan-1-yl acetate, do3aet, acmc-1bq9u, cyclen-n,n',n'-triacetic acid triethyl ester, 1,4,7,10-tetraazacyclododecane-1,4,7-triacetic acid triethyl ester, n,n',n'-tris-ethoxycarbonylmethyl-1,4,7,10-tetraazacyclododecane, 1,4,7,10-tetraazacyclododecane-1,4,7-triaceticacid, 1,4,7-triethyl ester |
| InChI Key | BPEUICGMDOXPJJ-UHFFFAOYSA-N |
| SMILES | CCOC(=O)CN1CCNCCN(CCN(CC1)CC(=O)OCC)CC(=O)OCC |
| IUPAC Name | ethyl 2-[4,7-bis(2-ethoxy-2-oxoethyl)-1,4,7,10-tetrazacyclododec-1-yl]acetate |
| Molecular Weight (g/mol) | 430.546 |
| PubChem CID | 10906130 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?