missing translation for 'onlineSavingsMsg'
Learn More

1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane

Product Code. 15407625
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15407625

Brand: Thermo Scientific Alfa Aesar H26792.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 157599-02-5 | C16H32N8O4 | 400.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157599-02-5
Molecular Formula C16H32N8O4
Molecular Weight (g/mol) 400.48
MDL Number MFCD09263318
InChI Key FQIHLPGWBOBPSG-UHFFFAOYSA-N
Synonym 2,2',2,2'-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetrayl tetraacetamide, dotam, 1,4,7,10-tetrakis aminocarbonylmethyl-1,4,7,10-tetraazacyclododecane, 2-4,7,10-tris carbamoylmethyl-1,4,7,10-tetraazacyclododecan-1-yl acetamide, acmc-209dgc, cyclen-n,n',n',n-tetraacetamide, 1,4,7,10-tetraazacyclododecane 1,4,7,10-tetraacetamide, 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetamide, 1,4,7,10-tetrakis carbamoylmethyl-1,4,7,10-tetraazacyclododecane
PubChem CID 11560395
IUPAC Name 2-[4,7,10-tris(2-amino-2-oxoethyl)-1,4,7,10-tetrazacyclododec-1-yl]acetamide
SMILES NC(=O)CN1CCN(CC(N)=O)CCN(CC(N)=O)CCN(CC(N)=O)CC1

Specifications

Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 400.48
Chemical Name or Material 1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.