missing translation for 'onlineSavingsMsg'
Learn More

1-(4,5-Dihydro-2-thiazolyl)-3-azetidinethiol Hydrochloride, TRC

Product Code. 30381004
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381004 250 mg
30369768 1 g
30382742 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381004 Supplier Toronto Research Chemicals Supplier No. TRCD454190250MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179337-57-6
Molecular Formula C6H11ClN2S2
Molecular Weight (g/mol) 210.75
Synonym 1-(4,5-Dihydro-2-thiazolyl)-3-azetidinethiol Monohydrochloride, 2-(3-Mercaptoazetidin-1-yl)-2-thiazoline Hydrochloride, 3-Mercapto-1-(1,3-thiazolin-2-yl)azetidine Hydrochloride
IUPAC Name 1-(4,5-dihydro-1,3-thiazol-2-yl)azetidine-3-thiol;hydrochloride
SMILES Cl.SC1CN(C1)C2=NCCS2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C6H10N2S2.ClH/c9-5-3-8(4-5)6-7-1-2-10-6;/h5,9H,1-4H2;1H
Formula Weight 210.0052
Chemical Name or Material 1-(4,5-Dihydro-2-thiazolyl)-3-azetidinethiol Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.