missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(4-hydroxyphenyl)piperazino]ethan-1-one, Thermo Scientific™

Product Code. p-4391365
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
This item is not returnable. View return policy

Product Code. 10795853

Brand: Thermo Scientific Maybridge SB02047EE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67914-60-7
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.27
MDL Number MFCD00044905
InChI Key AGVNLFCRZULMKK-UHFFFAOYSA-N
Synonym 1-acetyl-4-4-hydroxyphenyl piperazine, 4-1-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, n-acetyl-4-4-hydroxyphenyl piperazine, 4-4-acetyl-1-piperazinyl phenol, piperazine, 1-acetyl-4-4-hydroxyphenyl, 4-4-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethan-1-one, 1-acetyl-4-4-hydroxylphenyl piperazine
PubChem CID 712441
IUPAC Name 1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C1=CC=C(O)C=C1

Specifications

Assay Percent Range 97%
Quantity 50 g
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.