missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(4-hydroxyphenyl)piperazino]ethan-1-one, Thermo Scientific™

Product Code. 10557864
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10557864 100 g 100g
10795853 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10557864 Supplier Thermo Scientific Maybridge Supplier No. SB02047FA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67914-60-7
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.27
MDL Number MFCD00044905
InChI Key AGVNLFCRZULMKK-UHFFFAOYSA-N
Synonym 1-acetyl-4-4-hydroxyphenyl piperazine, 4-1-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, n-acetyl-4-4-hydroxyphenyl piperazine, 4-4-acetyl-1-piperazinyl phenol, piperazine, 1-acetyl-4-4-hydroxyphenyl, 4-4-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethan-1-one, 1-acetyl-4-4-hydroxylphenyl piperazine
PubChem CID 712441
IUPAC Name 1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C1=CC=C(O)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 100 g
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.