missing translation for 'onlineSavingsMsg'
Learn More

1-(4-(4-Hydroxyphenyl)butan-2-yl)-1H-indole-5,6-diol, TRC

Product Code. 30389957
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389957 5 mg 5mg
30397458 50 mg 50mg
30405360 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389957 Supplier Toronto Research Chemicals Supplier No. TRCH8256505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(4-(4-Hydroxyphenyl)butan-2-yl)-1H-indole-5,6-diol
Analyte or Component Names 1-(4-(4-Hydroxyphenyl)butan-2-yl)-1H-indole-5,6-diol
Shipping Condition Dry ice
Molecular Formula C18 H19 N O3
Synonym Dobutamine Aminochrome Impurity
InChI Formula InChI=1S/C18H19NO3/c1-12(2-3-13-4-6-15(20)7-5-13)19-9-8-14-10-17(21)18(22)11-16(14)19/h4-12,20-22H,2-3H2,1H3
Solution Type Neat
SMILES CC(CCc1ccc(O)cc1)n2ccc3cc(O)c(O)cc23
IUPAC Name 1-[4-(4-hydroxyphenyl)butan-2-yl]indole-5,6-diol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 297.348
Quantity 5 mg
Formula Weight 297.136
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.