missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine, 97%, Thermo Scientific™

Product Code. 10367114
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367114 5 g Amber Glass Bottle 5g
10361583 250 mg Amber Glass Bottle 250mg
10600915 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10367114 Supplier Thermo Scientific Maybridge Supplier No. CC33939DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 852227-96-4
Molecular Formula C17H26BNO2
Molecular Weight (g/mol) 287.21
MDL Number MFCD07368524
InChI Key OTOKWHGMHAAFRM-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 4-1-piperidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-piperidine, piperidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl, amtb222, 4-piperidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl piperidine
PubChem CID 4961250
IUPAC Name 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 287.21
Percent Purity 97%
Chemical Name or Material 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.