missing translation for 'onlineSavingsMsg'
Learn More

[1-[4-[4-(1,1-Dimethylethyl)-phenyl]but-3-enyl]piperidin-4-yl]diphenylmethanol, Mikromol™

Product Code. 30050946
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050946 25 mg 25mg
30050629 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050946 Supplier Mikromol Supplier No. MM0055.090025
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[1-[4-[4-(1,1-Dimethylethyl)-phenyl]but-3-enyl]piperidin-4-yl]diphenylmethanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1-(4-(4-(1,1-Dimethylethyl)-phenyl)but-3-enyl)piperidin-4-yl)diphenylmethanol
CAS 104953-06-2
Analyte or Component Names (1-(4-(4-(1,1-Dimethylethyl)-phenyl)but-3-enyl)piperidin-4-yl)diphenylmethanol
Shipping Condition Room Temperature
Molecular Formula C32 H39 N O
Certifications/Compliance ISO 17025
Synonym (1-[4-[4-(1,1-Dimethylethyl)-phenyl]but-3-enyl]piperidin-4-yl)diphenylmethanol, Terfenadine Imp. G (EP), Terfenadine Impurity G
InChI Formula InChI=1S/C32H39NO/c1-31(2,3)27-19-17-26(18-20-27)12-10-11-23-33-24-21-30(22-25-33)32(34,28-13-6-4-7-14-28)29-15-8-5-9-16-29/h4-10,12-20,30,34H,11,21-25H2,1-3H3/b12-10+
SMILES CC(C)(C)c1ccc(\C=C\CCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4)cc1
IUPAC Name [1-[(E)-4-(4-tert-butylphenyl)but-3-enyl]piperidin-4-yl]-diphenylmethanol
Molecular Weight (g/mol) 453.66
Quantity 25 mg
Formula Weight 453.3032 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.