missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(3-Bromothien-2-yl)phenyl]ethanone, 97%, Thermo Scientific™

Product Code. 10153624
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10153624 5 g Amber Glass Bottle 5g
10205434 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10153624 Supplier Thermo Scientific Maybridge Supplier No. CC67828DE
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 937796-01-5
Molecular Formula C12H9BrOS
Molecular Weight (g/mol) 281.167
MDL Number MFCD09879973
InChI Key AMADTVLVNSCDKW-UHFFFAOYSA-N
Synonym 1-4-3-bromothien-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethanone, 1-4-3-bromothiophen-2-yl phenyl ethan-1-one, 4'-3-bromothien-2-yl acetophenone
PubChem CID 24229756
IUPAC Name 1-[4-(3-bromothiophen-2-yl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)C2=C(C=CS2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 281.17
Percent Purity 97%
Chemical Name or Material 1-[4-(3-Bromothien-2-yl)phenyl]ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.