Learn More
Hamilton™ 1/4-28 Fittings and Washers (12 ga.) 10/pk
Product Code. 11522335
Change view
Click to view available options
Packaging Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. Packaging Quantity Unit Size
11522335 10/Pk. Pack of 10
1 options
Product Code. 11522335 Supplier Hamilton™ Supplier No. 88807
  • ONLINE EXCLUSIVE PRICE
    £127.00
    £117.38 / Pack of 10
    Save £9.62
    7% Off
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Hamilton offers a diverse line of inert fittings fittings and connectors made from CTFE. The fittings are designed for use with industry standard luer and luer lock products like our syringes as well as threaded 1/4-28 and M6 flat bottom ports.
Specifications
Type 1/4-28 Fittings and Washers (12 ga.) 10/pk
Packaging Quantity 10/Pk.