missing translation for 'onlineSavingsMsg'
Learn More

1-(4-(2-Hydroxy-3-(isopropylamino)propoxy)phenyl)-2-methoxyethanone, TRC

Product Code. 30393360
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393360 10 mg
30389669 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393360 Supplier Toronto Research Chemicals Supplier No. TRCH94366510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73723-85-0
Molecular Formula C15 H23 N O4
Molecular Weight (g/mol) 281.35
Synonym Ethanone, 1-[4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]-2-methoxy-
IUPAC Name 1-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]-2-methoxyethanone
SMILES COCC(=O)c1ccc(OCC(O)CNC(C)C)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H23NO4/c1-11(2)16-8-13(17)9-20-14-6-4-12(5-7-14)15(18)10-19-3/h4-7,11,13,16-17H,8-10H2,1-3H3
Formula Weight 281.1627
Chemical Name or Material 1-(4-(2-Hydroxy-3-(isopropylamino)propoxy)phenyl)-2-methoxyethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.