missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(1H-Indol-3-Yl)Piperidino]Ethan-1-One, TRC

Product Code. 30392174
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30392174 50 mg 50mg
30391345 500 mg -
30391637 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392174 Supplier Toronto Research Chemicals Supplier No. TRCI65584550MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30030-83-2
Molecular Formula C15H18N2O
Molecular Weight (g/mol) 242.31
Synonym 1-[4-(1H-Indol-3-yl)-1-piperidinyl]ethanone, 1-[4-(1H-Indol-3-yl)piperidin-1-yl]ethanone
IUPAC Name 1-[4-(1H-indol-3-yl)piperidin-1-yl]ethanone
SMILES CC(=O)N1CCC(CC1)c2c[nH]c3ccccc23

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H18N2O/c1-11(18)17-8-6-12(7-9-17)14-10-16-15-5-3-2-4-13(14)15/h2-5,10,12,16H,6-9H2,1H3
Formula Weight 242.1419
Chemical Name or Material 1-[4-(1H-Indol-3-Yl)Piperidino]Ethan-1-One
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.