missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(1H-Indol-3-Yl)Piperidino]Ethan-1-One, TRC

Product Code. 30391345
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
Product Code. Quantity unitSize
30391345 500 mg -
30391637 100 mg -
30392174 50 mg 50mg
3 options
This item is not returnable. View return policy

Product Code. 30391345

Brand: Toronto Research Chemicals TRCI655845500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30030-83-2
Molecular Formula C15H18N2O
Molecular Weight (g/mol) 242.31
Synonym 1-[4-(1H-Indol-3-yl)-1-piperidinyl]ethanone, 1-[4-(1H-Indol-3-yl)piperidin-1-yl]ethanone
IUPAC Name 1-[4-(1H-indol-3-yl)piperidin-1-yl]ethanone
SMILES CC(=O)N1CCC(CC1)c2c[nH]c3ccccc23

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H18N2O/c1-11(18)17-8-6-12(7-9-17)14-10-16-15-5-3-2-4-13(14)15/h2-5,10,12,16H,6-9H2,1H3
Formula Weight 242.1419
Chemical Name or Material 1-[4-(1H-Indol-3-Yl)Piperidino]Ethan-1-One
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.