missing translation for 'onlineSavingsMsg'
Learn More

1,3-Pyrenediamine, TRC

Product Code. 30415377
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415377 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415377 Supplier Toronto Research Chemicals Supplier No. TRCP98930050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Pyrenediamine
CAS 92821-64-2
Molecular Formula C16H12N2
Quantity 50 mg
Synonym 1,3-Diaminopyrene
InChI Formula InChI=1S/C16H12N2/c17-13-8-14(18)12-7-5-10-3-1-2-9-4-6-11(13)16(12)15(9)10/h1-8H,17-18H2
SMILES Nc1cc(N)c2ccc3cccc4ccc1c2c34
IUPAC Name pyrene-1,3-diamine
Molecular Weight (g/mol) 232.28
Formula Weight 232.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.