missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanesultone, TRC

Product Code. 30134839
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134839 100 g 100g
30041289 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134839 Supplier Toronto Research Chemicals Supplier No. TRCP760800100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-71-4
Molecular Formula C3 H6 O3 S
Molecular Weight (g/mol) 122.14
Synonym 1-Propanesulfonic acid, 3-hydroxy-, γ-sultone (6CI), 1,3-Propane sultone, 1,3-Trimethylene sultone, 3-Hydroxy-1-propanesulfonic acid sultone, 3-Hydroxy-1-propanesulfonic acid γ-sultone, NSC 42386, Propane sultone, Propyl sultone, γ-Propane sultone
IUPAC Name oxathiolane 2,2-dioxide
SMILES O=S1(=O)CCCO1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C3H6O3S/c4-7(5)3-1-2-6-7/h1-3H2
Formula Weight 122.0038
Chemical Name or Material 1,3-Propanesultone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.