missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Propanesultone, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.
Solubility
Soluble in water (100 mg/ml), chloroform, esters, methanol, and aromatic hydrocartions.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.
Solubility
Soluble in water (100 mg/ml), chloroform, esters, methanol, and aromatic hydrocartions.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
Specifications
Melting Point | 30°C to 33°C |
Density | 1.392 |
Boiling Point | 179°C to 181°C (30 mm) |
Flash Point | >110°C (230°F) |
Assay Percent Range | 99% |
Quantity | 50 g |
Beilstein | 109782 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water (100mg/ml),chloroform,esters,methanol,and aromatic hydrocartions. |
IUPAC Name | 1,2λ⁶-oxathiolane-2,2-dione |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'