missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanesultone, 99%

Product Code. 11408990
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408990 250 g 250g
11488980 10 g 10g
11498980 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408990 Supplier Thermo Scientific Alfa Aesar Supplier No. A11923.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1120-71-4 | C3H6O3S | 122.14 g/mol

1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.

Solubility
Soluble in water (100 mg/ml), chloroform, esters
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-71-4
Molecular Formula C3H6O3S
Molecular Weight (g/mol) 122.14
MDL Number MFCD00005355
InChI Key FSSPGSAQUIYDCN-UHFFFAOYSA-N
Synonym 1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9
PubChem CID 14264
ChEBI CHEBI:82370
SMILES O=S1(=O)CCCO1

Specifications

Melting Point 30°C to 33°C
Density 1.392
Boiling Point 179°C to 181°C (30 mmHg)
Flash Point >110°C (230°F)
Quantity 250 g
UN Number UN2811
Beilstein 109782
Sensitivity Moisture sensitive
Solubility Information Soluble in water (100mg/ml),chloroform,esters,methanol,and aromatic hydrocartions.
IUPAC Name oxathiolane 2,2-dioxide
Formula Weight 122.14
Percent Purity 99%
Chemical Name or Material 1,3-Propanesultone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.