missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanediol, 99%

Product Code. 11429782
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429782 5000 g 5000g
11419782 250 g 250g
11409782 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429782 Supplier Thermo Scientific Alfa Aesar Supplier No. A10829.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 504-63-2 | C3H8O2 | 76.10 g/mol

1,3-Propanediol is used as a solvent for thin film preparations, in the production of polymers such as polytrimethylene terephthalate, adhesives, laminates, coatings, moldings, aliphatic polyesters, as an antifreeze and in wood paint. It also acts as a reagent for vinyl epoxide synthon, for epoxide ring-opening, for polymerization reactions and for natural product syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Propanediol is used as a solvent for thin film preparations, in the production of polymers such as polytrimethylene terephthalate, adhesives, laminates, coatings, moldings, aliphatic polyesters, as an antifreeze and in wood paint. It also acts as a reagent for vinyl epoxide synthon, for epoxide ring-opening, for polymerization reactions and for natural product syntheses.

Solubility
Miscible with water and alcohol.

Notes
Incompatible with acid chlorides, acid anhydrides, oxidizing agents, chloroformates and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-63-2
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
MDL Number MFCD00002949
InChI Key YPFDHNVEDLHUCE-UHFFFAOYSA-N
Synonym 1,3-propanediol, trimethylene glycol, 1,3-dihydroxypropane, 2-deoxyglycerol, 1,3-propylene glycol, 1,3-propylenediol, 1,3-propandiol, 2-hydroxymethyl ethanol, omega-propanediol, beta-propylene glycol
PubChem CID 10442
ChEBI CHEBI:16109
IUPAC Name propane-1,3-diol
SMILES OCCCO

Specifications

Melting Point -26°C
Density 1.053
Boiling Point 214°C
Flash Point 140°C (174°F)
Refractive Index 1.44
Quantity 5000 g
Beilstein 969155
Merck Index 14,9714
Solubility Information Miscible with water and alcohol.
Formula Weight 76.1
Percent Purity 99%
Chemical Name or Material 1,3-Propanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.