missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propane sultone, 97%

Product Code. p-7077156 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
This item is not returnable. View return policy

Product Code. 15384968

Brand: Thermo Scientific Acros 131420250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1120-71-4 | C3H6O3S | 122.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-71-4
Molecular Formula C3H6O3S
Molecular Weight (g/mol) 122.14
MDL Number MFCD00005355
InChI Key FSSPGSAQUIYDCN-UHFFFAOYSA-N
Synonym 1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9
PubChem CID 14264
ChEBI CHEBI:82370
IUPAC Name oxathiolane 2,2-dioxide
SMILES O=S1(=O)CCCO1

Specifications

Melting Point 30.0°C to 33.0°C
Density 1.3920g/mL
Boiling Point 180.0°C (30.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 1.392
Formula Weight 122.14
Percent Purity 97%
Chemical Name or Material 1,3-Propane sultone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Propane sultone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,3-Propane sultone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.