missing translation for 'onlineSavingsMsg'
Learn More

1,3-Phenylenediacetic acid, 97%

Product Code. 11448790
Change view
Click to view available options
Quantity:
2 g
10 g
25 g
Unit Size:
10g
25g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448790 10 g 10g
11438790 2 g 2g
11490273 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448790 Supplier Thermo Scientific Alfa Aesar Supplier No. A11429.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19806-17-8 | C10H10O4 | 194.19 g/mol

In order to establish defined biomacromolecular systems for mineralised tissue applications, type I collagen was functionalised with 1,3-phenylenediacetic acid (Ph) and investigated at the molecular, macroscopic and functional levels. The solvothermal reactions of 1,2-bis(imidazol-1?-yl)ethane (bime), 1,3-phenylenediacetic acid, 1,4-phenylenediacetic acid), 1,2-phenylenediacetic acid with different metal ions gave rise to four novel helical coordination polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In order to establish defined biomacromolecular systems for mineralised tissue applications, type I collagen was functionalised with 1,3-phenylenediacetic acid (Ph) and investigated at the molecular, macroscopic and functional levels. The solvothermal reactions of 1,2-bis(imidazol-1′-yl)ethane (bime), 1,3-phenylenediacetic acid, 1,4-phenylenediacetic acid), 1,2-phenylenediacetic acid with different metal ions gave rise to four novel helical coordination polymers.

Solubility
Slightly soluble in water. Solublitiy in methanol is almost transparency.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19806-17-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004341
InChI Key GDYYIJNDPMFMTB-UHFFFAOYSA-N
Synonym 2,2'-1,3-phenylene diacetic acid, 1,3-phenylenediacetic acid, m-benzenediacetic acid, m-phenylenediacetic acid, 1,3-benzenediacetic acid, acetic acid, 2,2'-m-phenylene di, 2-3-carboxymethyl phenyl acetic acid, 1,3-benzenediaceticacid, 3-carboxymethyl phenyl acetic acid, 1,3-benzoldiessigsaure
PubChem CID 29788
IUPAC Name 2-[3-(carboxymethyl)phenyl]acetic acid
SMILES OC(=O)CC1=CC(CC(O)=O)=CC=C1

Specifications

Melting Point 172°C to 178°C
Odor Odorless
Quantity 10 g
Beilstein 2616749
Solubility Information Slightly soluble in water. Solublitiy in methanol is almost transparency.
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 1,3-Phenylenediacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.