missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Methoxyphenyl)piperazine dihydrochloride, 97%, Thermo Scientific™

Product Code. p-7043698
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311537 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11311537 Supplier Thermo Scientific Alfa Aesar Supplier No. L02164.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-(3-Methoxyphenyl)piperazine dihydrochloride is used as an arylpiperazine compound which is an important intermediate in the preparation of various pharmaceuticals, used to treat depression and post-traumatic stress.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6968-76-9
Molecular Formula C11H18Cl2N2O
Molecular Weight (g/mol) 265.178
MDL Number MFCD00035285
InChI Key UKUNKQNESKRETR-UHFFFAOYSA-N
Synonym 1-3-methoxyphenyl piperazine dihydrochloride, unii-q0t82qhg7n, 1-3-methoxyphenyl piperazine 2hcl, q0t82qhg7n, piperazine, 1-3-methoxyphenyl-, dihydrochloride, 1-3-methoxyphenyl piperazindihydrochlorid, 1-3-methoxyphenyl piperazine hcl, pubchem15280, 1-3-methoxyphenyl piperazinehydrochloride, n-3-methoxyphenyl piperazine dihydrochloride
PubChem CID 81429
IUPAC Name 1-(3-methoxyphenyl)piperazine;dihydrochloride
SMILES COC1=CC=CC(=C1)N2CCNCC2.Cl.Cl

Specifications

Melting Point ∽214°C (decomposition)
Quantity 25 g
Beilstein 5647713
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 265.18
Percent Purity 97%
Chemical Name or Material 1-(3-Methoxyphenyl)piperazine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.