missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Methoxyphenyl)piperazine, 95%

Product Code. 15421698
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 15421698

Brand: Thermo Scientific Alfa Aesar H55450.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16015-71-7 | C11H16N2O | 192.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16015-71-7
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD00040733
InChI Key PZIBVWUXWNYTNL-UHFFFAOYSA-N
Synonym 1-3-methoxyphenyl piperazine, n-3-methoxyphenyl piperazine, 1-3-methoxyphenyl-piperazine, unii-6lcp52962g, 1-3-methoxy-phenyl-piperazine, chembl59597, piperazine, 1-3-methoxyphenyl, 3-methoxy-1-piperazinylbenzene, pubchem8585, 3-meopp
PubChem CID 81430
IUPAC Name 1-(3-methoxyphenyl)piperazine
SMILES COC1=CC=CC(=C1)N2CCNCC2

Specifications

Density 1.114
Boiling Point 150°C (0.5 mmHg)
Flash Point 91°C (196°F)
Refractive Index 1.5815
Quantity 25 g
UN Number UN3267
Formula Weight 192.26
Percent Purity 95%
Chemical Name or Material 1-(3-Methoxyphenyl)piperazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.