missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Isocyanatophenyl)-1H-pyrrole, ≥97%, Thermo Scientific™

Product Code. 10473223
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-59-1
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.198
MDL Number MFCD07772799
InChI Key PUPFPJGLDBUOOI-UHFFFAOYSA-N
Synonym 1-3-isocyanatophenyl-1h-pyrrole, 1-3-isocyanatophenyl pyrrole, 3-pyrrolylbenzenisocyanate, 1h-pyrrole,1-3-isocyanatophenyl
PubChem CID 7162333
IUPAC Name 1-(3-isocyanatophenyl)pyrrole
SMILES C1=CN(C=C1)C2=CC=CC(=C2)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 184.2
Percent Purity 97%
Chemical Name or Material 1-(3-Isocyanatophenyl)-1H-pyrrole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'