missing translation for 'onlineSavingsMsg'
Learn More

1,3-Indanedione, 97%

Product Code. 11474873
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474873 25 g 25g
11464873 5 g 5g
11484873 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474873 Supplier Thermo Scientific Alfa Aesar Supplier No. A12206.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 606-23-5 | C9H6O2 | 146.145 g/mol

It is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.

Solubility
Soluble in ethanol, ether, benzene, slightly soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-23-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.145
MDL Number MFCD00003779
InChI Key UHKAJLSKXBADFT-UHFFFAOYSA-N
Synonym 1,3-indanedione, 1,3-indandione, 1h-indene-1,3 2h-dione, 1,3-diketohydrindene, indan-1,3-dione, 1,3-indanone, indane-1,3-dione, diketohydrindene, 1,3-indandion, unii-4djn7yg35g
PubChem CID 11815
ChEBI CHEBI:78877
IUPAC Name indene-1,3-dione
SMILES C1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 128°C to 132°C
Density 1.37
Quantity 25 g
Beilstein 1210061
Solubility Information Soluble in ethanol,ether,benzene,slightly soluble in water.
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material 1,3-Indanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.