missing translation for 'onlineSavingsMsg'
Learn More

1,3-Indandione, TRC

Product Code. 30402346
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402346 10 g 10g
30396417 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402346 Supplier Toronto Research Chemicals Supplier No. TRCI50800510G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Indanedione
CAS 606-23-5
Molecular Formula C9 H6 O2
Quantity 10 g
Synonym 1H-Indene-1,3(2H)-dione (9CI, ACI), 1,3-Indandione (8CI), Indandione (6CI), 1,3-Diketohydrindene, 1,3-Dioxobenzocyclopentane, 1,3-Indanedione, 2H-Indene-1,3-dione, Indene-1,3(2H)-dione, NSC 26329, NSC 6312
InChI Formula InChI=1S/C9H6O2/c10-8-5-9(11)7-4-2-1-3-6(7)8/h1-4H,5H2
SMILES O=C1CC(=O)c2ccccc12
IUPAC Name indene-1,3-dione
Molecular Weight (g/mol) 146.14
Formula Weight 146.0368
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.