missing translation for 'onlineSavingsMsg'
Learn More

1,3-Glyceryl Dilinoleate (>80%), TRC

Product Code. 30393813
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393813 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393813 Supplier Toronto Research Chemicals Supplier No. TRCG60152010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Glyceryl Dilinoleate
CAS 15818-46-9
Molecular Formula C39 H68 O5
Quantity 10 mg
Synonym (Z,Z)-9,12-Octadecadienoic Acid 2-Hydroxy-1,3-propanediyl Ester, α,α'-Dilinolein, 1,3-Dilinoleoyl-rac-glycerol, (Z,Z)-9,12-Octadecadienoic Acid 2-Hydroxy-1,3-propanediyl Ester
InChI Formula InChI=1S/C39H68O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-35-37(40)36-44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h11-14,17-20,37,40H,3-10,15-16,21-36H2,1-2H3/b13-11-,14-12-,19-17-,20-18-
SMILES CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC(O)COC(=O)CCCCCCC\C=C/C\C=C/CCCCC
IUPAC Name [2-hydroxy-3-[(9Z,12Z)-octadeca-9,12-dienoyl]oxypropyl] (9Z,12Z)-octadeca-9,12-dienoate
Molecular Weight (g/mol) 616.95
Formula Weight 616.5067
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.