missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Fluorophenyl)-2-thiourea, 98%, Thermo Scientific™

Product Code. 10198742 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10198742 1 g Glass bottle 1g
10495023 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10198742 Supplier Thermo Scientific Acros Supplier No. 372510010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 458-05-9
Molecular Formula C7H7FN2S
Molecular Weight (g/mol) 170.21
InChI Key YKOBLHQALWQKTC-UHFFFAOYSA-N
Synonym 1-3-fluorophenyl-2-thiourea, 1-3-fluorophenyl thiourea, 3-fluorophenyl thiourea, n-3-fluorophenyl thiourea, amino 3-fluorophenyl amino methane-1-thione, acmc-20apbn, 1-m-fluorophenyl thiourea, 3-fluoro-phenyl-thiourea
PubChem CID 2734799
IUPAC Name (3-fluorophenyl)thiourea
SMILES C1=CC(=CC(=C1)F)NC(=S)N

Specifications

Melting Point 113°C to 117°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula F6C6H4NHCSNH2
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 170.21
Percent Purity 98%
Chemical Name or Material 1-(3-Fluorophenyl)-2-thiourea
compliance-icons
Product Identifier
  • 1-(3-Fluorophenyl)-2-thiourea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.