missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Fluorophenyl)-2-thiourea, 98%, Thermo Scientific™

Product Code. p-4812541 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10198742

Brand: Thermo Scientific Acros 372510010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 458-05-9
Molecular Formula C7H7FN2S
Molecular Weight (g/mol) 170.21
InChI Key YKOBLHQALWQKTC-UHFFFAOYSA-N
Synonym 1-3-fluorophenyl-2-thiourea, 1-3-fluorophenyl thiourea, 3-fluorophenyl thiourea, n-3-fluorophenyl thiourea, amino 3-fluorophenyl amino methane-1-thione, acmc-20apbn, 1-m-fluorophenyl thiourea, 3-fluoro-phenyl-thiourea
PubChem CID 2734799
IUPAC Name (3-fluorophenyl)thiourea
SMILES C1=CC(=CC(=C1)F)NC(=S)N

Specifications

Melting Point 113°C to 117°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula F6C6H4NHCSNH2
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 170.21
Percent Purity 98%
Chemical Name or Material 1-(3-Fluorophenyl)-2-thiourea
compliance-icons
Product Identifier
  • 1-(3-Fluorophenyl)-2-thiourea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.