missing translation for 'onlineSavingsMsg'
Learn More

1,3-Divinylbenzene (Stabilized with TBC), TRC

Product Code. 30138646
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138646 1 g 1g
30133740 500 mg 500mg
30005855 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138646 Supplier Toronto Research Chemicals Supplier No. TRCD6794131G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-57-6
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
Synonym 1,3-Diethenylbenzene, 1,3-Bis(ethenyl)benzene, 1,3-Divinylbenzene, 3-Vinylstyrene, m-Divinylbenzene, m-Vinylstyrene
SMILES C=Cc1cccc(C=C)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H10/c1-3-9-6-5-7-10(4-2)8-9/h3-8H,1-2H2
Formula Weight 130.0783
Chemical Name or Material 1,3-Divinylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.