missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dithiolan-2-imine, TRC

Product Code. 30380288
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380288 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380288 Supplier Toronto Research Chemicals Supplier No. TRCD49469525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dithiolan-2-imine
CAS 4472-81-5
Molecular Formula C3H5NS2
Quantity 25 mg
Synonym Dithioimidocarbonic Acid Cyclic Ethylene Ester, 1,2-Ethanedithiol Cyclic Dithioimidocarbonate, 1,2-Ethanedithiol Cyclic Ester With Dithioimidocarbonic Acid, Carbonimidodithioic Acid Cyclic 1,2-Ethanediyl Ester
InChI Formula InChI=1S/C3H5NS2/c4-3-5-1-2-6-3/h4H,1-2H2
SMILES N=C1SCCS1
IUPAC Name 1,3-dithiolan-2-imine
Molecular Weight (g/mol) 119.21
Formula Weight 118.9863
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.