missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dithietan-2-imine Hydrochloride, TRC

Product Code. 30384045
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384045 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384045 Supplier Toronto Research Chemicals Supplier No. TRCD49240510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dithietan-2-imine Hydrochloride
CAS 25878-14-2
Molecular Formula C2H3NS2.HCl
Quantity 10 mg
Synonym Dithio-imidocarbonic Acid Cyclic Methylene Ester Hydrochloride, Methanedithiol Cyclic Dithioimidocarbonate Hydrochloride, 2-Imino-1,3-dithietane Hydrochloride
InChI Formula InChI=1S/C2H3NS2.ClH/c3-2-4-1-5-2;/h3H,1H2;1H
SMILES Cl.N=C1SCS1
IUPAC Name 1,3-dithietan-2-imine;hydrochloride
Molecular Weight (g/mol) 141.6
Formula Weight 140.9474
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.