missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dipropyl-8-phenylxanthine, Thermo Scientific™

Product Code. 15415759
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415759 10 mg 10mg
15425759 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415759 Supplier Thermo Scientific Alfa Aesar Supplier No. J63554.MA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective A1 adenosine antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85872-53-3
Molecular Formula C17H20N4O2
Molecular Weight (g/mol) 312.373
MDL Number MFCD00209811
InChI Key CLIGSMOZKDCDRZ-UHFFFAOYSA-N
Synonym 1,3-dipropyl-8-phenylxanthine, 3,7-dihydro-1,3-dipropyl-8-phenyl-1h-purine-2,6,dione, 8-phenyl-1,3-dipropyl-9h-purine-2,6-dione, 8-phenyl-1,3-dipropyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6,dione, 3,7-dihydro-1,3-dipropyl-8-phenyl, 1h-purine-2,6-dione, 3,7-dihydro-8-phenyl-1,3-dipropyl, tocris-0486, 8-phenyl-1,3-dipropyl-3,9-dihydro-1h-purine-2,6-dione, 8-phenyl-1,3-dipropyl-3,9-dihydro-purine-2,6-dione
PubChem CID 128784
IUPAC Name 8-phenyl-1,3-dipropyl-7H-purine-2,6-dione
SMILES CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C3=CC=CC=C3

Specifications

Color White
Quantity 10 mg
Solubility Information Soluble in ethanol (5 mM), or DMSO (10 mM) with warming
Formula Weight 312.37
Physical Form Powder
Chemical Name or Material 1,3-Dipropyl-8-phenylxanthine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.