missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylguanidine, TRC

Product Code. 18071330
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18071330 2.5 g 2.5g
18053130 500 mg 500mg
18041730 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18071330 Supplier Toronto Research Chemicals Supplier No. TRCD4626732.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-06-7
Molecular Formula C13 H13 N3
Molecular Weight (g/mol) 211.26
Synonym Guanidine, N,N'-diphenyl-, Guanidine, 1,3-diphenyl- (8CI), N,N'-Diphenylguanidine, 1,2-Diphenylguanidine, 1,3-Diphenylguanidine, Accel D, Accel DM-R, Accelerator BR, Accelerator D, Accelerator DPG, Chlorostain BR, DFG, DPG, DPG 75, DPG 80, Denax, Denax DPG, Diphenylguanidine, Guanid F, Melaniline, NSC 3272, Nocceler D, Nocceler D-P, Oricel D, Perkacit DP, Perkacit DPG, Rhenocure DPG, Rhenogran DPG, Rhenogran DPG 80, Rhenogran DPG 80P, Sanceler D, Sanceler D-G, Soxinol D, Soxinol DG, Vanax DPG, Vulcafor DPG, Vulkacit D, Vulkacit D/C, Vulkacit D/EG-C, Vulkazit, sym-Diphenylguanidine
IUPAC Name 1,3-diphenylguanidine
SMILES N=C(Nc1ccccc1)Nc2ccccc2

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C13H13N3/c14-13(15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H,(H3,14,15,16)
Formula Weight 211.1109
Chemical Name or Material 1,3-Diphenylguanidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.