missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyl-2-propyn-1-ol, tech. 90%

Product Code. p-7035655
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15430128

Brand: Thermo Scientific Alfa Aesar H53374.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1817-49-8 | C15H12O | 208.26 g/mol

1,3-Diphenyl-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenyl-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1817-49-8
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD06654198
InChI Key DZZWMODRWHHWFR-UHFFFAOYSA-N
Synonym 1,3-diphenyl-2-propyn-1-ol, 1,3-diphenyl-prop-2-yn-1-ol, 1-phenyl-3-phenyl-propyn-3-ol, 1,3-diphenylpropargyl alcohol, benzenemethanol, a-2-phenylethynyl, alpha-phenyl ethynyl-benzenemethanol
PubChem CID 296659
IUPAC Name 1,3-diphenylprop-2-yn-1-ol
SMILES C1=CC=C(C=C1)C#CC(C2=CC=CC=C2)O

Specifications

Density 1.099
Flash Point >110°C (230°F)
Refractive Index 1.618
Quantity 1 g
UN Number UN3082
Formula Weight 208.26
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,3-Diphenyl-2-propyn-1-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.