missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyl-2-propyn-1-ol, tech. 90%

Product Code. 15440128
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440128 5 g 5g
15430128 1 g 1g
15450128 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440128 Supplier Thermo Scientific Alfa Aesar Supplier No. H53374.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1817-49-8 | C15H12O | 208.26 g/mol

1,3-Diphenyl-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenyl-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1817-49-8
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD06654198
InChI Key DZZWMODRWHHWFR-UHFFFAOYSA-N
Synonym 1,3-diphenyl-2-propyn-1-ol, 1,3-diphenyl-prop-2-yn-1-ol, 1-phenyl-3-phenyl-propyn-3-ol, 1,3-diphenylpropargyl alcohol, benzenemethanol, a-2-phenylethynyl, alpha-phenyl ethynyl-benzenemethanol
PubChem CID 296659
IUPAC Name 1,3-diphenylprop-2-yn-1-ol
SMILES C1=CC=C(C=C1)C#CC(C2=CC=CC=C2)O

Specifications

Density 1.099
Flash Point >110°C (230°F)
Refractive Index 1.618
Quantity 5 g
UN Number UN3082
Formula Weight 208.26
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,3-Diphenyl-2-propyn-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.