missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyl-1-butanone, 95%, Thermo Scientific Chemicals

Product Code. p-7025262
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469097 1 g 1g
11479097 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469097 Supplier Thermo Scientific Alfa Aesar Supplier No. B20654.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1533-20-6 | Molecular Formula: C16H16O | Molecular Weight: 224.303 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1533-20-6
Molecular Formula C16H16O
Molecular Weight (g/mol) 224.303
MDL Number MFCD00026345
InChI Key GIVFXLVPKFXTCU-UHFFFAOYSA-N
Synonym 1,3-diphenyl-1-butanone, 3-phenylbutyrophenone, b-phenylbutyrophenone, acmc-20ap3x, 1,3-diphenyl-butan-1-one, phenyl 2-phenylpropyl ketone, 1-butanone, 1,3-diphenyl, 2-alpha-methylbenzyl acetophenone
PubChem CID 137065
IUPAC Name 1,3-diphenylbutan-1-one
SMILES CC(CC(=O)C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 68°C to 72°C
Quantity 1 g
Formula Weight 224.3
Percent Purity 95%
Chemical Name or Material 1,3-Diphenyl-1-butanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.