missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyl-1,3-propanedione, 98+%

Product Code. 11445513
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445513 25 g 25g
11455513 100 g 100g
11465513 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445513 Supplier Thermo Scientific Alfa Aesar Supplier No. A13450.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-46-7 | C15H12O2 | 224.259 g/mol

1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.

Solubility
It is Soluble in ether, chloroform, and aqueous sodium hydroxide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Acid anhydrides, Halogens, halogen acids. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-46-7
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00003085
InChI Key NZZIMKJIVMHWJC-UHFFFAOYSA-N
Synonym dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
PubChem CID 8433
ChEBI CHEBI:75417
IUPAC Name 1,3-diphenylpropane-1,3-dione
SMILES C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2

Specifications

Melting Point 76°C to 80°C
Boiling Point 219°C to 221°C (18 mmHg)
Quantity 25 g
Beilstein 514910
Merck Index 14,3009
Solubility Information It is Soluble in ether,chloroform,and aqueous sodium hydroxide. Insoluble in water.
Formula Weight 224.26
Percent Purity ≥98%
Chemical Name or Material 1,3-Diphenyl-1,3-propanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.