missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenoxybenzene, 99%

Product Code. 10652075 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10652075 5 g Glass bottle 5g
10548322 25 g Glass bottle 25g
11904721 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10652075 Supplier Thermo Scientific Acros Supplier No. 408840050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3379-38-2 | C18H14O2 | 262.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3379-38-2
Molecular Formula C18H14O2
Molecular Weight (g/mol) 262.31
MDL Number MFCD00003036
InChI Key JTNRGGLCSLZOOQ-UHFFFAOYSA-N
Synonym m-diphenoxybenzene, benzene, 1,3-diphenoxy, m-diphenyloxybenzene, m-phenoxyphenoxybenzene, benzene, m-diphenoxy, resorcinol diphenyl ether, 1,1'-1,3-phenylenebis oxy dibenzene, m-diphenoxybenzol, acmc-1adxk
PubChem CID 76911
ChEBI CHEBI:39273
IUPAC Name 1,3-diphenoxybenzene
SMILES O(C1=CC=CC=C1)C1=CC(OC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 58.0°C to 61.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 262.3
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1, 3-Diphenoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.