missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenoxybenzene, 99%, Thermo Scientific™

Product Code. 11477977
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477977 5 g 5g
11487977 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477977 Supplier Thermo Scientific Alfa Aesar Supplier No. A16647.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3379-38-2
Molecular Formula C18H14O2
Molecular Weight (g/mol) 262.31
MDL Number MFCD00003036
InChI Key JTNRGGLCSLZOOQ-UHFFFAOYSA-N
Synonym m-diphenoxybenzene, benzene, 1,3-diphenoxy, m-diphenyloxybenzene, m-phenoxyphenoxybenzene, benzene, m-diphenoxy, resorcinol diphenyl ether, 1,1'-1,3-phenylenebis oxy dibenzene, m-diphenoxybenzol, acmc-1adxk
PubChem CID 76911
ChEBI CHEBI:39273
IUPAC Name 1,3-diphenoxybenzene
SMILES O(C1=CC=CC=C1)C1=CC(OC2=CC=CC=C2)=CC=C1

Especificaciones

Melting Point 59°C to 61°C
Quantity 5 g
Beilstein 2054179
Formula Weight 262.31
Percent Purity 99%
Chemical Name or Material 1,3-Diphenoxybenzene

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.