missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, ReagentPlus™, 99%, Honeywell™

Product Code. 15621660 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Aluminum Bottle
Glass Bottle
Unit Size:
1 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15621660 500 mL Glass Bottle 500mL
15611660 1 L Glass Bottle 1 L
15608330 4 L Aluminum Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15621660 Supplier Honeywell Research Chemicals Supplier No. 184497500ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, contains ∼75 ppm BHT as inhibitor, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-06-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00003207
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 12586
ChEBI CHEBI:87597
IUPAC Name 1,3-dioxolane
SMILES C1COCO1

Specifications

Melting Point -95°C
Color Colorless
Density 1.06g/mL at 25°C
Boiling Point 76°C
Flash Point −3°C (26.6°F)
Ignition Point 525°F
Packaging Glass Bottle
Refractive Index n20/D 1.401(lit.)
Quantity 500 mL
UN Number UN1166
Beilstein 102453
Formula Weight 74.08g/mol
Vapor Pressure 70mmHg (20°C)
Percent Purity 99%
Grade ReagentPlus
Chemical Name or Material 1,3-Dioxolane, Contains approx.75 ppm BHT as inhibitor
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.