missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, 99.5%, stab.

Product Code. 11330148
Change view
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
Unit Size:
1 L
250mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11330148 1 L 1 L
11350148 250 mL 250mL
11340148 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11330148 Supplier Thermo Scientific Alfa Aesar Supplier No. 042897.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As a solvent, in Mannich reaction, as an electrolyte in batteries | CAS: 646-06-0 | C3H6O2 | 74.079 g/mol

As a solvent, in Mannich reaction, as an electrolyte in batteries.1,3-Dioxolane is used as a solvent for paints, cellulose esters and in organic synthesis. It is used as an extragent for waxes, fats and oils, stabilizer for halogenated hydrocarbons and co monomer in polyacetals preparation. It is used as finishing agent and swelling agent in textiles. It acts as an intermediate for the preparation of acyclovir and vandetanib. It is an essential ingredient in industrial polymers. In polymerization reactions, it acts as a chain length regulator and a chain transfer agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-06-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00003207
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 12586
ChEBI CHEBI:87597
IUPAC Name 1,3-dioxolane
SMILES C1COCO1

Specifications

Melting Point -95°C
Density 1.06 g/mL
Boiling Point 74°C to 75°C
Flash Point 1°C (34°F)
Refractive Index 1.4
Quantity 1 L
UN Number UN1166
Solubility Information Miscible with water,ethanol,ether,n-octanol,acetone,chloroform,dichloromethane,ethyl Acetate and methanol.
Formula Weight 74.08
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material 1,3-Dioxolane, Stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.